Please login to your online account to display your discounted pricing

Hydroxymethylpolystyrene resin, 1% crossl. with DVB, 1.0 to 1.2mmol/g, 100 to 200 mesh


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  349510050

 View more versions of this product

Catalog No. AC349510050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 5g
Chemical Name or Material Hydroxymethylpolystyrene resin
Packaging Glass bottle
CAS 66072-40-0

Specifications

Chemical Name or Material Hydroxymethylpolystyrene resin
CAS 66072-40-0