Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydroxymethylferrocene, 97%

$471.01 - $1,517.73

Chemical Identifiers

Chemical Name or Material Hydroxymethylferrocene
CAS 1273-86-5
Formula Weight 216.07
MDL Number MFCD00040878
Synonym Ferrocenemethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3925706

 
alfa aesar™
3925706
5g Each for $471.01

AA3925714

 
alfa aesar™
3925714
25g Each for $1,517.73
Description & Specifications

Specifications

Formula Weight 216.07
Chemical Name or Material Hydroxymethylferrocene
Physical Form Crystalline
Assay 97%
MDL Number MFCD00040878
CAS 1273-86-5
Synonym Ferrocenemethanol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Moisture Sensitive
Color Yellow
Melting Point 79° to 81°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.