Please login to your online account to display your discounted pricing

Alfa Aesar™ (Hydroxymethyl)triphenylphosphonium chloride, 98%

$721.16 - $721.16

Chemical Identifiers

Chemical Name or Material (Hydroxymethyl)triphenylphosphonium chloride
CAS 5293-83-4
Molecular Formula HOCH2P(C6H5)3Cl
Formula Weight 328.78g/mol
MDL Number MFCD00031688
Beilstein 4170800
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1861530

 
alfa aesar™
A18615-30
250g Each for $721.16
Description & Specifications

Specifications

Melting Point 194° to 198°C
Quantity 250g
Chemical Name or Material (Hydroxymethyl)triphenylphosphonium chloride
CAS 5293-83-4
Assay 98%
Molecular Formula HOCH2P(C6H5)3Cl
Formula Weight 328.78g/mol
MDL Number MFCD00031688
Beilstein 4170800
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.