Learn More
Hydroxymethanesulfinic acid sodium salt hydrate, tech 85%, cont. up to 5% sodium sulfite (dry wt.), Thermo Scientific Chemicals
$61.96 - $235.16
Chemical Identifiers
CAS | 149-44-0 |
---|---|
Molecular Formula | CH3NaO3S |
Molecular Weight (g/mol) | 118.08 |
MDL Number | MFCD00040426 |
InChI Key | XWGJFPHUCFXLBL-UHFFFAOYSA-M |
Synonym | sodium hydroxymethanesulfinate, rongalite, rongalit, aldanil, discolite, formapon, formopan, hydrolit, leptacid, leptacit |
PubChem CID | 23689980 |
SMILES | [Na+].OCS([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1573136
|
Thermo Scientific Chemicals
A1573136 |
500 g |
Each for $61.96
|
|
|||||
AAA157310E
|
Thermo Scientific Chemicals
A157310E |
2500 g |
Each for $235.16
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
149-44-0 | |
118.08 | |
XWGJFPHUCFXLBL-UHFFFAOYSA-M | |
23689980 |
CH3NaO3S | |
MFCD00040426 | |
sodium hydroxymethanesulfinate, rongalite, rongalit, aldanil, discolite, formapon, formopan, hydrolit, leptacid, leptacit | |
[Na+].OCS([O-])=O |
Specifications
149-44-0 | |
CH3NaO3S | |
500 g | |
14,8620 | |
XWGJFPHUCFXLBL-UHFFFAOYSA-M | |
sodium;hydroxymethanesulfinate | |
23689980 | |
85% | |
Hydroxymethanesulfinic acid sodium salt hydrate |
64°C to 68°C | |
MFCD00040426 | |
3630814 | |
sodium hydroxymethanesulfinate, rongalite, rongalit, aldanil, discolite, formapon, formopan, hydrolit, leptacid, leptacit | |
[Na+].OCS([O-])=O | |
118.08 | |
118.09 (Anhydrous) | |
Technical | |
cont. up to 5% sodium sulfite (dry wt.) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-739-4
RTECSNumber : PB0380000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only