Learn More
Hydroxymethanesulfinic acid sodium salt hydrate, tech 85%, cont. up to 5% sodium sulfite (dry wt.), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A157310E
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hydroxymethanesulfinic acid sodium salt hydrate | |
64°C to 68°C | |
CH3NaO3S | |
2500 g | |
14,8620 | |
XWGJFPHUCFXLBL-UHFFFAOYSA-M | |
sodium hydroxymethanesulfinate | |
23689980 | |
85% |
149-44-0 | |
cont. up to 5% sodium sulfite (dry wt.) | |
MFCD00040426 | |
3630814 | |
sodium hydroxymethanesulfinate, rongalite, rongalit, aldanil, discolite, formapon, formopan, hydrolit, leptacid, leptacit | |
[Na+].OCS([O-])=O | |
118.08 | |
118.09 (Anhydrous) | |
Technical |
Chemical Identifiers
149-44-0 | |
118.08 | |
XWGJFPHUCFXLBL-UHFFFAOYSA-M | |
23689980 |
CH3NaO3S | |
MFCD00040426 | |
sodium hydroxymethanesulfinate, rongalite, rongalit, aldanil, discolite, formapon, formopan, hydrolit, leptacid, leptacit | |
[Na+].OCS([O-])=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-739-4
RTECSNumber : PB0380000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only