missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydroxymethanesulfinic acid sodium salt hydrate, tech 85%, cont. up to 5% sodium sulfite (dry wt.)

Manufacturer:  Alfa Aesar™ A1573136

 View more versions of this product

Catalog No. AAA1573136



Specifications

Specifications

Hydroxymethanesulfinic acid sodium salt hydrate
500g
cont. up to 5% sodium sulfite (dry wt.)
MFCD00150598
14,8620
XWGJFPHUCFXLBL-UHFFFAOYSA-M
sodium;hydroxymethanesulfinate
23689980
85%
64°C to 68°C
149-44-0
CH3NaO3S
3630814
sodium hydroxymethanesulfinate, rongalite, rongalit, aldanil, discolite, formapon, formopan, hydrolit, leptacid, leptacit
C(O)S(=O)[O-].[Na+]
118.082
118.09 (Anhydrous)
Technical

Chemical Identifiers

149-44-0
118.082
XWGJFPHUCFXLBL-UHFFFAOYSA-M
23689980
C(O)S(=O)[O-].[Na+]
CH3NaO3S
MFCD00150598
sodium hydroxymethanesulfinate, rongalite, rongalit, aldanil, discolite, formapon, formopan, hydrolit, leptacid, leptacit
sodium;hydroxymethanesulfinate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : PB0380000

TSCA : TSCA

Documents