missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Hydroxylamine Sulfate (Crystalline/Certified ACS), Fisher Chemical™
Manufacturer: Fisher Chemical FLH331100
Specifications
Hydroxylamine Sulfate | |
Glass Bottle | |
170°C | |
3.6 | |
≥99 % | |
(NH2OH)2·H2SO4 | |
Oxammonium Sulfate | |
Solid | |
Certified |
Pass Test | |
White | |
100g | |
10039-54-0 | |
H6N2O2 · H2SO4 | |
MFCD00044869 | |
164.14 | |
≥99% |
Chemical Identifiers
10039-54-0 | |
H6N2O2 · H2SO4 | |
MFCD00044869 | |
Oxammonium Sulfate |
Safety and Handling

DOTInformation : DOT Class 8, : Corrosive