missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroxylamine Sulfate (Crystalline/Certified ACS), Fisher Chemical

$150.20 - $263.46

Chemical Identifiers

CAS 10039-54-0
Molecular Formula H6N2O2 · H2SO4
MDL Number MFCD00044869
Synonym Oxammonium Sulfate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
H331-100 SDS View Documents Fisher Chemical
FLH331100
100g Glass Bottle Each for $150.20
Case of 6 Each for $901.20
 
H331-500 SDS View Documents Fisher Chemical
H331500
500g Glass Bottle Each for $263.46
Case of 6 Each for $1,580.76
 
Specifications

Chemical Identifiers

10039-54-0
H6N2O2 · H2SO4
MFCD00044869
Oxammonium Sulfate

Specifications

Pass Test
White
100g
10039-54-0
H6N2O2 · H2SO4
MFCD00044869
164.14
≥99%
Hydroxylamine Sulfate
Glass Bottle
170°C
3.6
≥99 %
(NH2OH)2·H2SO4
Oxammonium Sulfate
Solid
Certified
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive