missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroxylamine Sulfate (Crystalline/Certified ACS), Fisher Chemical™

Manufacturer:  Fisher Chemical H331500

 View more versions of this product

Catalog No. H331-500




Specifications

Specifications

Hydroxylamine Sulfate
Pass Test
White
500g
10039-54-0
H6N2O2 · H2SO4
MFCD00044869
Oxammonium Sulfate
Solid
Certified
1.7g/mL
Glass Bottle
170°C
3.6
≥99 %
(NH2OH)2·H2SO4
2865
164.14
≥99%

Chemical Identifiers

10039-54-0
H6N2O2 · H2SO4
MFCD00044869
Oxammonium Sulfate
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents