Please login to your online account to display your discounted pricing

Hydroxylamine sulfate, 99%, ACROS Organics™


Quantity 5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC198530051

 View more versions of this product

Catalog No. AC198530051

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystals
Assay ≥98.5%
Formula Weight 164.14
Quantity 5kg
Color White
Fieser 01,481
Merck Index 15, 4867
Infrared Spectrum Authentic
Packaging Plastic bottle
Molecular Formula H6N2O2·H2SO4
Linear Formula (H2NOH)2·H2SO4
CAS 10039-54-0
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
May be corrosive to metals.
Suspected of causing cancer if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life.
Chemical Name or Material Hydroxylamine sulfate
Assay Percent Range 99%
MDL Number MFCD00044869
Synonym Hydroxylammonium sulfate
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Hydroxylamine sulfate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula H6N2O2·H2SO4
Linear Formula (H2NOH)2·H2SO4
Formula Weight 164.14
MDL Number MFCD00044869
Synonym Hydroxylammonium sulfate
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
May be corrosive to metals.
Suspected of causing cancer if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life.

SDS & Safety Documents