Please login to your online account to display your discounted pricing

Hydroxylamine Hydrochloride, Hygroscopic, ACS Grade, ≥ 96.0% , LabChem™

$204.97 - $204.97

Chemical Identifiers

Chemical Name or Material Hydroxylamine Hydrochloride
CAS 5470-11-1
CAS Max % 100
Name Note Hygroscopic
Purity Grade ACS Grade
Linear Formula H3NO·HCl
Formula Weight 69.49
UN Number UN2923
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC155151

 
lab chem
LC155151
Poly Bottle 500g Each for $204.97
Description & Specifications

Specifications

Formula Weight 69.49
Assay ≥96.0%
Health Hazard 2 GHS H Statement
May be corrosive to metals.
Harmful if swallowed or in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing cancer.
May cause damage to organs (blood) through prolonged or repeated exposure.
Very toxic to aquatic life.
Solubility Information Soluble in water
Specific Gravity 1670kg/m3
Purity Grade ACS Grade
Decomposition Information Gaseous Ammonia; Hydrogen chloride
CAS 5470-11-1
Color Colorless
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep only in original container.
Do not breathe dust.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Contaminated work clothing mustd not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Acidity ≤0.25meq/gcSt
Chemical Name or Material Hydroxylamine Hydrochloride
Melting Point 152°C
Quantity 500g
Health Hazard 1 Warning
Linear Formula H3NO·HCl
Identification Passes Test
Physical Form Solid
EINECS Number 226-798-2
Density 1,670kg/m3
Name Note Hygroscopic
Packaging Poly Bottle
Storage Information Room Temperature
UN Number UN2923
CAS Max % 100
Safety and Handling

Warning

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep only in original container.
Do not breathe dust.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Contaminated work clothing mustd not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May be corrosive to metals.
Harmful if swallowed or in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing cancer.
May cause damage to organs (blood) through prolonged or repeated exposure.
Very toxic to aquatic life.