Please login to your online account to display your discounted pricing

Hydroxylamine Hydrochloride, For Calcium, Magnesium, 3% (w/v), LabChem™

$80.09 - $80.09

Chemical Identifiers

Chemical Name or Material Hydroxylamine Hydrochloride
CAS 7732-18-5
CAS Max % 97
CAS 5470-11-1
CAS Max % 3
Name Note For Calcium, Magnesium
Linear Formula H3NO·HCl
Formula Weight 69.49
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC155201

 
lab chem
LC155201
500mL Each for $80.09
Description & Specifications

Specifications

Formula Weight 69.49
Storage Information Room Temperature
Identification Passes Test
Solubility Soluble in water
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash skin with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS 7732-18-5
Physical Form Liquid
Quantity 500mL
Assay 3% (w/v)
Linear Formula H3NO·HCl
CAS 5470-11-1
Packaging Poly Bottle
Decomposition Information Gaseous Ammonia; Carbon monoxide; Carbon dioxide; Nitrogen oxides
Health Hazard 2 GHS H Statement
May cause an allergic skin reaction.
Suspected of causing cancer.
May cause damage to organs (blood) through prolonged or repeated exposure.
Toxic to aquatic life.
Color Clear, colorless
Health Hazard 1 Warning
CAS Max % 97
Chemical Name or Material Hydroxylamine Hydrochloride
Name Note For Calcium, Magnesium
CAS Max % 3
Safety and Handling

Warning

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash skin with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May cause an allergic skin reaction.
Suspected of causing cancer.
May cause damage to organs (blood) through prolonged or repeated exposure.
Toxic to aquatic life.