Please login to your online account to display your discounted pricing

Hydroxylamine Hydrochloride, For Cadmium, 20% (w/v), LabChem™

$101.77 - $101.77

Chemical Identifiers

Chemical Name or Material Hydroxylamine Hydrochloride
CAS 7732-18-5
CAS Max % 80
CAS 5470-11-1
CAS Max % 20
Name Note For Cadmium
Linear Formula H3NO·HCl
Formula Weight 69.49
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC155401

 
lab chem
LC155401
500mL Each for $101.77
Description & Specifications

Specifications

Formula Weight 69.49
Solubility Soluble in water
Color Clear, colorless
Physical Form Liquid
Identification Passes Test
Name Note For Cadmium
Storage Information Room Temperature
Linear Formula H3NO·HCl
Packaging Poly Bottle
CAS 7732-18-5
Decomposition Information Gaseous Ammonia; Nitrogen oxides; Hydrogen chloride
Assay 20% (w/v)
Health Hazard 2 GHS H Statement
Harmful if swallowed or in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing cancer.
May cause damage to organs (blood) through prolonged or repeated exposure.
Toxic to aquatic life.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash skin with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Chemical Name or Material Hydroxylamine Hydrochloride
CAS Max % 20
Quantity 500mL
CAS 5470-11-1
Health Hazard 1 Warning
CAS Max % 80
Safety and Handling

Warning

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash skin with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed or in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing cancer.
May cause damage to organs (blood) through prolonged or repeated exposure.
Toxic to aquatic life.