Please login to your online account to display your discounted pricing

Hydroxylamine hydrochloride, ACS reagent, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  412051000

 View more versions of this product

Catalog No. AC412051000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Purity Grade ACS Reagent Grade
Physical Form Crystalline Powder, Crystals, and/or Chunks
Formula Weight 69.49
Quantity 100g
Color White
Merck Index 15, 4867
EINECS Number 226-798-2
TSCA TSCA
RTECS Number NC3675000
Fieser 01,478; 02,217; 07,176; 09,245
CAS 1304222
Infrared Spectrum Authentic
Iron (Fe) 5ppm max.
Packaging Plastic bottle
Ammonium Passes Test
Residue after Ignition 0.05% max.
Sulfur Compounds 0.005% max.
Titratable Free Acid 0.25 meq/g max.
Synonym Oxammonium hydrochloride
Clarity Passes Test
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Heavy Metals (as Pb) 5ppm max.
Chemical Name or Material Hydroxylamine hydrochloride
Name Note 96+%
Molecular Formula H3NO·HCl
Linear Formula H2NOH·HCl
MDL Number MFCD00051089
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause an allergic skin reaction.
Harmful if swallowed.
Very toxic to aquatic life.
Harmful in contac

Specifications

Chemical Name or Material Hydroxylamine hydrochloride
Name Note 96+%
CAS 1304222
Purity Grade ACS Reagent Grade
Assay ≥96.0%
Molecular Formula H3NO·HCl
Linear Formula H2NOH·HCl
Formula Weight 69.49
MDL Number MFCD00051089
Synonym Oxammonium hydrochloride
Accessories

Warning

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause an allergic skin reaction.
Harmful if swallowed.
Very toxic to aquatic life.
Harmful in contac

SDS & Safety Documents