missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroxylamine hydrochloride, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 270102500

 View more versions of this product

Catalog No. AC270102500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

Hydroxylamine hydrochloride
5470-11-1
Crystalline Powder, Crystals and/or Chunks
99+%
H2NOH·HCl
01,478; 02,217; 07,176; 09,245
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
WTDHULULXKLSOZ-UHFFFAOYSA-N
hydroxylamine;hydrochloride
443297
69.49
Authentic
155°C to 158°C
White
250 g
ClH4NO
MFCD00051089
15, 4867
Solubility in water: 560g/L (20°C). Other solubilities: soluble in alcohol,methanol,glycerol,propylene glycol,insoluble in cold ether
Cl.NO
69.49
CHEBI:5807
≥99%
Plastic bottle

Chemical Identifiers

5470-11-1
69.49
WTDHULULXKLSOZ-UHFFFAOYSA-N
443297
Cl.NO
ClH4NO
MFCD00051089
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
CHEBI:5807
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing canc

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protecti

GHS Signal Word: Danger

EINECSNumber : 226-798-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only