missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroxylamine hydrochloride, 99+%, Thermo Scientific™

Manufacturer:  Fisher Scientific Acros 270101000

 View more versions of this product

Catalog No. AC270101000



Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Reduction reagent
Specifications

Specifications

Hydroxylamine hydrochloride
99+%
H2NOH·HCl
01,478; 02,217; 07,176; 09,245
hydroxyamine hcl, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine hcl, hydroxylamine hydrochloride, hydroxylamine, hydrochloride, hydroxylamine, hydrochloride 1:1, hydroxylammonium chloride, oxammonium hci, oxammonium hydrochloride
WTDHULULXKLSOZ-UHFFFAOYSA-N
hydroxylamine;hydrochloride
443297
69.49
99+%
0.001% max.
0.005% max
155°C to 158°C
5470-11-1
H3NO·HCl
MFCD00051089
15, 4867
Solubility in water: 560g/L (20°C). Other solubilities: soluble in alcohol,methanol,glycerol,propylene glycol,insoluble in cold ether
NO.Cl
69.49
CHEBI:5807
Crystalline Powder, Crystals, and/or Chunks
Authentic
Plastic bottle
White
100g

Chemical Identifiers

5470-11-1
69.49
WTDHULULXKLSOZ-UHFFFAOYSA-N
443297
hydroxylamine;hydrochloride
H3NO·HCl
MFCD00051089
hydroxyamine hcl, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine hcl, hydroxylamine hydrochloride, hydroxylamine, hydrochloride, hydroxylamine, hydrochloride 1:1, hydroxylammonium chloride, oxammonium hci, oxammonium hydrochloride
CHEBI:5807
NO.Cl
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing canc

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protecti

GHS Signal Word: Danger

EINECSNumber : 226-798-2

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only