Please login to your online account to display your discounted pricing

Hydroxylamine-EDTA, For Selenium, LabChem™

$86.40 - $86.40

Chemical Identifiers

Chemical Name or Material Hydroxylamine-EDTA
CAS 7732-18-5
CAS Max % 96.6
CAS 5470-11-1
CAS Max % 2.5
CAS 6381-92-6
CAS Max % 0.9
Name Note For Selenium
Linear Formula H3NO·HCl
Formula Weight 69.49
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC155101

 
lab chem
LC155101
500mL Each for $86.40
Description & Specifications

Specifications

Formula Weight 69.49
Chemical Name or Material Hydroxylamine-EDTA
CAS Max % 0.9
Decomposition Information Gaseous Ammonia; Carbon monoxide; Carbon dioxide; Nitrogen oxides
Color Clear, colorless
Quantity 500mL
Name Note For Selenium
Physical Form Liquid
Linear Formula H3NO·HCl
CAS 7732-18-5
Health Hazard 1 Warning
CAS Max % 96.6
Health Hazard 2 GHS H Statement
May cause an allergic skin reaction.
Suspected of causing cancer.
May cause damage to organs (blood) through prolonged or repeated exposure.
Toxic to aquatic life.
Identification Passes Test
CAS 5470-11-1
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 2.5
Solubility Soluble in water
CAS 6381-92-6
Storage Information Room Temperature
Safety and Handling

Warning

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May cause an allergic skin reaction.
Suspected of causing cancer.
May cause damage to organs (blood) through prolonged or repeated exposure.
Toxic to aquatic life.