Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydroxyapatite Nanoparticles, 5-10% (w/v) aqueous colloidal dispersion


20-50nm particles

Manufacturer: alfa aesar™  J66691AE

 View more versions of this product

Catalog No. AAJ66691AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 100mL
Format Liquid dispersion
Storage Requirements 2°C to 8°C
CAS 12167-74-7
Molecular Formula Ca5HO13P3
Formula Weight 502.31
Synonym nHAP; Nano- hydroxyapatite
Melting Point 1100°