Please login to your online account to display your discounted pricing

Hydroquinone sulfonic acid, potassium salt, tech., ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  354681000

 View more versions of this product

Catalog No. AC354681000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Physical Form Powder
Formula Weight 228.25
Quantity 100g
Color Off-white to gray
Infrared Spectrum Authentic
MDL Number MFCD00007475
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Packaging Plastic bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Hydroquinone sulfonic acid, potassium salt
Linear Formula (HO)2C6H3SO3K
CAS 21799-87-1
Name Note tech.
Molecular Formula C6H5KO5S

Specifications

Chemical Name or Material Hydroquinone sulfonic acid, potassium salt
Name Note tech.
CAS 21799-87-1
Purity Grade Technical Grade
Molecular Formula C6H5KO5S
Linear Formula (HO)2C6H3SO3K
Formula Weight 228.25
MDL Number MFCD00007475
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents