Please login to your online account to display your discounted pricing

Hydroquinone sulfonic acid, potassium salt, tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material Hydroquinone sulfonic acid, potassium salt
Name Note tech.
CAS 21799-87-1
Purity Grade Technical Grade
Molecular Formula C6H5KO5S
Linear Formula (HO)2C6H3SO3K
Formula Weight 228.25
MDL Number MFCD00007475
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC354681000

  SDS
thermo fisher scientific
354681000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC354680050

  SDS
thermo fisher scientific
354680050
Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Technical Grade
Physical Form Powder
Formula Weight 228.25
Quantity 100g
Color Off-white to gray
Infrared Spectrum Authentic
Packaging Plastic bottle
MDL Number MFCD00007475
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Hydroquinone sulfonic acid, potassium salt
Name Note tech.
Molecular Formula C6H5KO5S
Linear Formula (HO)2C6H3SO3K
CAS 21799-87-1
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.