Please login to your online account to display your discounted pricing

Hydroquinone sulfonic acid, potassium salt, tech., ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  354680050

 View more versions of this product

Catalog No. AC354680050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass Bottle
Color Off-white to gray
Quantity 5g
Chemical Name or Material Hydroquinone sulfonic acid, potassium salt
Name Note tech.
CAS 21799-87-1
Purity Grade Technical
Physical Form Off-white to gray powder
Molecular Formula C6H5KO5S
Linear Formula (HO)2C6H3SO3K
Formula Weight 228.25g/mol
MDL Number MFCD00007475
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Hydroquinone sulfonic acid, potassium salt
Name Note tech.
CAS 21799-87-1
Purity Grade Technical
Molecular Formula C6H5KO5S
Linear Formula (HO)2C6H3SO3K
Formula Weight 228.25g/mol
MDL Number MFCD00007475
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents