Please login to your online account to display your discounted pricing

Hydroquinone, Reagent Grade, LabChem™

$142.06 - $142.06

Chemical Identifiers

Chemical Name or Material Hydroquinone
CAS 123-31-9
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula 1,4-HOC6H4OH
Formula Weight 110.11
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC154901

 
lab chem
LC154901
Amber Glass 500g Each for $142.06
Description & Specifications

Specifications

Formula Weight 110.11
Chemical Name or Material Hydroquinone
EINECS Number 204-617-8
Density 1.32g/cm3
Identification Passes Test
Purity Grade Reagent Grade
Physical Form Powder
Linear Formula 1,4-HOC6H4OH
Packaging Amber Glass
CAS 123-31-9
UN Number UN3077
Solubility Information Soluble in water
Health Hazard 1 Danger
CAS Max % 100
Health Hazard 2 GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
Suspected of causing genetic defects.
Suspected of causing cancer.
Very toxic to aquatic life.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Specific Gravity 1.32g/cm3
Recommended Storage Room Temperature
Decomposition Information Carbon monoxide; Carbon dioxide
Flash Point 165°C
Color White
Quantity 500g
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
Suspected of causing genetic defects.
Suspected of causing cancer.
Very toxic to aquatic life.

Room Temperature