Please login to your online account to display your discounted pricing

Hydroquinone-O,O'-diacetic acid, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  346030050

 View more versions of this product

Catalog No. AC346030050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Color Off-white to light tan
Quantity 5g
Chemical Name or Material Hydroquinone-O, O';-diacetic acid
CAS 2245-53-6
Physical Form Off-white to light tan powder
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C10H10O6
Linear Formula C6H4(OCH2COOH)2
Formula Weight 226.19g/mol
MDL Number MFCD00016816
Beilstein 06, IV, 5746
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 218-834-0
TSCA TSCA

Specifications

Chemical Name or Material Hydroquinone-O, O';-diacetic acid
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C10H10O6
Linear Formula C6H4(OCH2COOH)2
Formula Weight 226.19g/mol
MDL Number MFCD00016816
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents