Please login to your online account to display your discounted pricing

Hydroquinone-O,O'-diacetic acid, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Hydroquinone-O, O';-diacetic acid
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C10H10O6
Linear Formula C6H4(OCH2COOH)2
Formula Weight 226.19
MDL Number MFCD00016816
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC346030250

  SDS
thermo fisher scientific
346030250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 226.19
Quantity 25g
Color Off-white to light tan
Beilstein 06, IV, 5746
EINECS Number 218-834-0
TSCA TSCA
Chemical Name or Material Hydroquinone-O, O';-diacetic acid
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C10H10O6
Linear Formula C6H4(OCH2COOH)2
Packaging Glass bottle
MDL Number MFCD00016816
CAS 2245-53-6
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.