missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroquinone, For Chloride, Certified, 1% (w/v), LabChem™

$69.34 - $69.34

Chemical Identifiers

CAS 7732-18-5,123-31-9
CAS Max % 99,1
Linear Formula 1,4-HOC6H4OH
Formula Weight 110.11
Chemical Name or Material Hydroquinone, For Chloride
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC155001 Lab Chem
LC155001
500mL Each for $69.34
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,123-31-9
1,4-HOC6H4OH
Hydroquinone, For Chloride
99,1
110.11
Certified Grade

Specifications

7732-18-5,123-31-9
Liquid
110.11
Amber Glass
Room Temperature
Clear to slightly peach
Hydroquinone, For Chloride
Certified Grade
99,1
1,4-HOC6H4OH
Passes Test
Soluble in water
Carbon monoxide; Carbon dioxide
500mL
1% (w/v)

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful to aquatic life.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing mist.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Warning