missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydroquinone bis(2-hydroxyethyl)ether, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 218221000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Hydroquinone bis(2-hydroxyethyl)ether | |
104-38-1 | |
Authentic | |
Plastic bottle | |
C6H4(OCH2CH2OH)2 | |
100g | |
hydroquinone bis 2-hydroxyethyl ether, 1,4-bis 2-hydroxyethoxy benzene, vernatzer 30/10, 2,2'-1,4-phenylenebis oxy diethanol, hydroquinone di 2-hydroxyethyl ether, hydroquinone diethylol ether, 2,2'-phenylenedioxy diethanol, 2,2'-p-phenylenedioxy diethanol, 2,2'-p-phenylenedioxydiethanol, 2-4-2-hydroxyethoxy phenoxy ethanol | |
WTPYFJNYAMXZJG-UHFFFAOYSA-N | |
2-[4-(2-hydroxyethoxy)phenoxy]ethanol | |
66912 | |
99+% |
99+% | |
185.0°C to 200.0°C (0.3mmHg) | |
99% min. (GC) | |
C10H14O4 | |
MFCD00002861 | |
06, III, 4406 | |
Solubility in water: practically insoluble. Other solubilities: ca 4g/100mL ethanol,ca 4g/100mL acetone | |
C1=CC(=CC=C1OCCO)OCCO | |
198.22 | |
198.22 |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 203-197-3
TSCA : TSCA
RUO â Research Use Only