missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroquinone, 99%, ACROS Organics™

$59.88 - $169.08

Chemical Identifiers

CAS 123-31-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00002339
InChI Key QIGBRXMKCJKVMJ-UHFFFAOYSA-N
Synonym 1,4-benzenediol, 1,4-dihydroxybenzene, 4-hydroxyphenol, benzoquinol, hydroquinone, p-benzenediol, p-dihydroxybenzene, p-hydroquinone, p-hydroxyphenol, quinol
PubChem CID 785
ChEBI CHEBI:17594
IUPAC Name benzene-1,4-diol
SMILES C1=CC(=CC=C1O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120915000
SDS View Documents
Thermo Fisher Scientific
120915000
500g Plastic bottle
Each for $59.88
 
AC120910020
SDS View Documents
Thermo Fisher Scientific
120910020
2kg Plastic bottle
Each for $169.08
 
Description

Description

Specifications

Chemical Identifiers

123-31-9
110.11
QIGBRXMKCJKVMJ-UHFFFAOYSA-N
785
benzene-1,4-diol
C6H6O2
MFCD00002339
1,4-benzenediol, 1,4-dihydroxybenzene, 4-hydroxyphenol, benzoquinol, hydroquinone, p-benzenediol, p-dihydroxybenzene, p-hydroquinone, p-hydroxyphenol, quinol
CHEBI:17594
C1=CC(=CC=C1O)O

Specifications

1.32
Authentic
285.0°C to 287.0°C
170.0°C to 174.0°C
123-31-9
C6H6O2
06, 836
15, 4845
Solubility in water: 70g/L in water (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene,readily soluble in ethanol,acetone and methanol
C1=CC(=CC=C1O)O
110.11
CHEBI:17594
Needle-Like Crystals or Crystalline Powder
Hydroquinone, 99%
165°C
Plastic bottle
White
500g
98.5% min. (HPLC)
MFCD00002339
05,341; 14,249
1,4-benzenediol, 1,4-dihydroxybenzene, 4-hydroxyphenol, benzoquinol, hydroquinone, p-benzenediol, p-dihydroxybenzene, p-hydroquinone, p-hydroxyphenol, quinol
QIGBRXMKCJKVMJ-UHFFFAOYSA-N
benzene-1,4-diol
785
110.11
99%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Suspected of causing cancer.
Harmful if swallowed.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-617-8

SDS