Learn More
Hydroquinone, 99.5%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 219930010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Hydroquinone | |
123-31-9 | |
285.0°C to 287.0°C | |
Authentic | |
Plastic bottle | |
C6H4(OH)2 | |
1kg | |
05,341; 14,249 | |
hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol | |
QIGBRXMKCJKVMJ-UHFFFAOYSA-N | |
benzene-1,4-diol | |
785 | |
110.11 |
p.a. | |
1.32 | |
165°C | |
99.4% min. (HPLC) | |
C6H6O2 | |
MFCD00002339 | |
06,836 | |
15,4845 | |
Solubility in water: 70g/L in water (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene,readily soluble in ethanol,acetone and methanol | |
C1=CC(=CC=C1O)O | |
110.11 | |
CHEBI:17594 | |
99.5% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Suspected of causing cancer.
Harmful if swallowed.
Very toxic to aquatic life.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 204-617-8
RTECSNumber : MX3500000
TSCA : TSCA
RUO â Research Use Only