missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroquinidine Hydrochloride, TCI America™

$197.09 - $1,098.50

Chemical Identifiers

CAS 1476-98-8
Molecular Formula C20H27ClN2O2
Molecular Weight (g/mol) 362.898
MDL Number MFCD00135598
InChI Key MULXTQKDWYBJMO-VJAUXQICSA-N
Synonym Dihydroquinidine Hydrochloride
PubChem CID 16211709
IUPAC Name (S)-[(2R,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;hydrochloride
SMILES CCC1CN2CCC1CC2C(C3=C4C=C(C=CC4=NC=C3)OC)O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H075225G View Documents TCI America
H075225G
25g Each for $197.09
 
H0752250G View Documents TCI America
H0752250G
250g Each for $1,098.50
 
Specifications

Chemical Identifiers

1476-98-8
362.898
MULXTQKDWYBJMO-VJAUXQICSA-N
16211709
CCC1CN2CCC1CC2C(C3=C4C=C(C=CC4=NC=C3)OC)O.Cl
C20H27ClN2O2
MFCD00135598
Dihydroquinidine Hydrochloride
(S)-[(2R,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;hydrochloride

Specifications

White
1476-98-8
MFCD00135598
MULXTQKDWYBJMO-VJAUXQICSA-N
(S)-[(2R,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;hydrochloride
16211709
Crystal-Powder at 20°C
Hydroquinidine Hydrochloride
25g
C20H27ClN2O2
Dihydroquinidine Hydrochloride
CCC1CN2CCC1CC2C(C3=C4C=C(C=CC4=NC=C3)OC)O.Cl
362.898
362.90
≥98.0% (T,HPLC)
Safety and Handling

Safety and Handling

RTECSNumber : MX3018000

TSCA : No