Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min

Chemical Identifiers

Chemical Name or Material Hydrogen tetrachloroaurate(III) hydrate
CAS 27988-77-8
Formula Weight 339.79 (Anhydrous)
MDL Number MFCD00149903
UN Number UN3260
Synonym Tetrachloroauric(III) acid; Chloroauric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1232506

  SDS
alfa aesar™
1232506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 339.79 (Anhydrous)
TSCA TSCA
CAS 27988-77-8
Health Hazard 1 Danger
Assay 99.9%
UN Number UN3260
Synonym Tetrachloroauric(III) acid; Chloroauric acid
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Light Sensitive & Hygroscopic
Health Hazard 2 GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Assay Percent Notes (metals basis)
RTECS Number MD5428000
Concentration Au 49% min.
Quantity 5g
MDL Number MFCD00149903
Chemical Name or Material Hydrogen tetrachloroaurate(III) hydrate
Color Yellow-orange
Physical Form Crystalline
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.