Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrogen tetrabromoaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 32% min

Chemical Identifiers

Chemical Name or Material Hydrogen tetrabromoaurate(III) hydrate
Purity Grade Premion™
Formula Weight 517.61 (Anhydrous)
MDL Number MFCD00054118
UN Number UN3260
Synonym Bromoauric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4043303

  SDS
alfa aesar™
4043303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4043306

  SDS
alfa aesar™
4043306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 517.61 (Anhydrous)
Sensitivity Hygroscopic
Physical Form Crystalline
MDL Number MFCD00054118
UN Number UN3260
Chemical Name or Material Hydrogen tetrabromoaurate(III) hydrate
Purity Grade Premion™
Quantity 1g
TSCA TSCA
Concentration Au 32% min.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Solubility Information Soluble in water and alcohol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Synonym Bromoauric acid
Assay 99.99%
Assay Percent Notes (metals basis)
Melting Point ≈27°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.