missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen Peroxide USP, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Formula Weight 34.01
Chemical Name or Material Hydrogen Peroxide, Topical Solution
Grade USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK52324 Avantor Performance
523210*
Glass 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7722-84-1
34.01
USP
H2O2
Hydrogen Peroxide, Topical Solution

Specifications

34.01
7722-84-1
Glass
USP
4L
H2O2
Hydrogen Peroxide, Topical Solution