Please login to your online account to display your discounted pricing

Hydrogen Peroxide, Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative, 2.5 to 3.5%, U.S.P., J.T.Baker™


Quantity 500mL
Packaging Narrow Mouth Amber Glass Bottle

Manufacturer: avantor performance  218201

 View more versions of this product

Catalog No. 02003086

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay 2.5 to 3.5%
Formula Weight 34.01
Chemical Name or Material Hydrogen Peroxide
Molecular Formula H2O2
CAS 7722-84-1
Name Note Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative
Purity Grade USP
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.00kg
Quantity 500mL

Specifications

Chemical Name or Material Hydrogen Peroxide
CAS 7722-84-1
Name Note Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative
Purity Grade USP
Molecular Formula H2O2
Formula Weight 34.01