Please login to your online account to display your discounted pricing

Hydrogen Peroxide Solution 3% AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Hydrogen Peroxide Solution
Assay Percent Range 3%
Molecular Formula H2O2
Formula Weight 34.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK5241500

 
avantor performance
524104
Glass 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK52418

 
avantor performance
524108
Glass 8PT CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 34.01
Quantity 500mL
Molecular Formula H2O2
Chemical Name or Material Hydrogen Peroxide Solution
Assay Percent Range 3%
CAS 7722-84-1
Packaging Glass