Please login to your online account to display your discounted pricing

Hydrogen Peroxide, Certified, 2.0% (w/w) to 2.5% (w/w), LabChem™

$64.46 - $64.46

Chemical Identifiers

Chemical Name or Material Hydrogen Peroxide
CAS 7732-18-5
CAS Max % 98
CAS 7722-84-1
CAS Max % 2
Purity Grade Certified Grade
Linear Formula H2O2
Formula Weight 34.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC154402

 
lab chem
LC154402
Poly Bottle 1L Each for $64.46
Description & Specifications

Specifications

Formula Weight 34.01
Assay 2.0 to 2.5% (w/w)
Purity Grade Certified Grade
Recommended Storage Room Temperature
Density 1g/mL
Health Hazard 1 Warning
Packaging Poly Bottle
Color Clear, colorless
Quantity 1L
Identification Passes Test
CAS Max % 98
Chemical Name or Material Hydrogen Peroxide
CAS 7722-84-1
CAS 7732-18-5
Linear Formula H2O2
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Decomposition Information Oxygen
Physical Form Liquid
Solubility Information Soluble in water
CAS Max % 2
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.

GHS H Statement
Causes skin irritation.
Causes serious eye damage.

Room Temperature