Learn More
Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab., Thermo Scientific Chemicals
For Bleaching and Deodorizing Textiles, Wood, Hair
Supplier: Thermo Scientific Chemicals 033323AY
Description
For bleaching and deodorizing textiles, wood, hair, etc; rocket propulsion; maturing agent in food; water and sewage treatment; disinfectant; laboratory reagentHydrogen peroxide is used as an oxidizing agent to prepare propylene oxide, dibenzoyl peroxide, lauryl peroxide, peracetic acid and meta-chloroperoxybenzoic acid. It is also involved in the production of inorganic chemicals like sodium perborate and sodium percarbonate, which are used as bleaching agents in detergents. It is an associated iron catalyst and utilized for waste-water treatment to remove organic impurities through an advanced oxidation process. It is also used as a laboratory disinfectant and as a topical antiseptic. In the cosmetic industry, it is used for bleaching human hair and tooth whitening by mixing with ammonium hydroxide and baking soda respectively. Monopropellant engines using hydrogen peroxide as the fuel are commonly used for air-independent underwater propulsion applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hydrogen peroxide | |
107°C | |
MFCD00011333 | |
3587191 | |
1.11 | |
Soluble in water,alcohol and ether. Insoluble in petroleumMiscible with water,ether,alcohol,organic solvents and carboxylic esters. Immiscible with hydrocarbons and petroleum ether. | |
OO | |
34.014 | |
CHEBI:16240 | |
ACS | |
1.335 | |
4 x 500 mL |
7722-84-1 | |
H2O2 | |
UN2014 | |
29-32% w/w aq. Soln | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
34.02 | |
Plastic bottle | |
1.11 | |
Liquid |
Chemical Identifiers
7722-84-1 | |
34.014 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |
H2O2 | |
MFCD00011333 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
Safety and Handling
P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P370+P378q-P501c
H271-H302+H332-H314
DOTInformation : Hazard Class: 5.1; Packaging Group: II
EINECSNumber : 231-765-0
RTECSNumber : MX0899000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only