missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrogen Peroxide, Certified, >30% (w/w), LabChem™
Supplier: LabChem LC154301
Specifications
Hydrogen Peroxide | |
7722-84-1,7732-18-5 | |
-0.41°C | |
H2O2 | |
UN2014 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
34.01 | |
Certified | |
Poly Bottle | |
Traceable to NIST | |
500 mL |
Colorless | |
150.2°C | |
67.5,32.5 | |
H2O2 | |
1.41g/cm3 | |
Soluble in water | |
OO | |
34.014 | |
CHEBI:16240 | |
>30% (w/w) | |
Passes Test | |
1.41g/cm3 | |
Oxygen; Thermal decomposition generates: Corrosive vapors | |
Liquid |
Chemical Identifiers
7722-84-1 | |
34.014 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
H2O2 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed or if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe mist, vapors, spray.
Danger
EINECSNumber : 231-765-0
Recommended Storage : Room Temperature