missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen Peroxide, 10% (w/w) Aqueous Solution Reagent Grade, Stabilized with Acetanilide, Ricca Chemical

$64.82 - $64.82

Chemical Identifiers

CAS 7722-84-1,7732-18-5
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide, 10% (w/w) Aqueous Solution, Stabilized with Acetanilide
Grade Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
R38213101BV Ricca Chemical Company
R38213101BV
Amber Plastic Bottle 1L Each for $64.82
R38213104BV Ricca Chemical Company
RIR38213104BV
Amber Plastic Bottle 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7722-84-1,7732-18-5
Liquid
Hydrogen Peroxide, 10% (w/w) Aqueous Solution, Stabilized with Acetanilide
Reagent Grade

Specifications

7722-84-1,7732-18-5
Amber Plastic Bottle
1L
Reagent Grade
Liquid
Clear colorless
Hydrogen Peroxide, 10% (w/w) Aqueous Solution, Stabilized with Acetanilide