Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrogen hexafluoroantimonate(V), ≈65% aq. soln.

Chemical Identifiers

Chemical Name or Material Hydrogen hexafluoroantimonate(V)
CAS 16950-06-4
Linear Formula HF·SbF5
Formula Weight 236.75
MDL Number MFCD00011327
UN Number UN3264
Synonym Hexafluoroantimonic acid; Fluoroantimonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3348814

  SDS
alfa aesar™
3348814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3348818

  SDS
alfa aesar™
3348818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3348822

  SDS
alfa aesar™
3348822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.75
Chemical Name or Material Hydrogen hexafluoroantimonate(V)
TSCA TSCA
Physical Form Liquid
Linear Formula HF·SbF5
MDL Number MFCD00011327
CAS 16950-06-4
UN Number UN3264
Synonym Hexafluoroantimonic acid; Fluoroantimonic acid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H302-H332
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Concentration ≈65% aq. soln.
Color Colorless to dark orange
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H302-H332
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.