Please login to your online account to display your discounted pricing

hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  195370010

 View more versions of this product

Catalog No. AC195370010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 409.82
Physical Form Powder and/or Chunks
Quantity 1g
Color Orange to orange-brown
Assay 37.5 to 40.5% (Pt)
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
CAS 26023-84-7
Name Note ca. 40% Pt
Molecular Formula H2Cl6Pt·xH2O
Chemical Name or Material Hydrogen hexachloroplatinate(IV) hydrate
Linear Formula H2PtCl6·xH2O
MDL Number MFCD00149909
Total Trace Metal Impurities 0.1% max.
Synonym Hexachloroplatinic acid
Packaging Glass bottle
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger

Specifications

Chemical Name or Material Hydrogen hexachloroplatinate(IV) hydrate
Name Note ca. 40% Pt
CAS 26023-84-7
Assay 37.5 to 40.5% (Pt)
Molecular Formula H2Cl6Pt·xH2O
Linear Formula H2PtCl6·xH2O
Formula Weight 409.82
MDL Number MFCD00149909
Synonym Hexachloroplatinic acid
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

SDS & Safety Documents