Learn More
hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 195370250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Hydrogen Hexachloroplatinate(IV) Hydrate | |
26023-84-7 | |
H2Cl6Pt·xH2O | |
MFCD00149909 | |
Hexachloroplatinic acid, Platinic chloride | |
409.82 | |
ca. 40% Pt |
ca. 40% Pt | |
25g | |
H2PtCl6·xH2O | |
UN 2507 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
409.82 | |
Glass Bottle |
Safety and Handling
GHS H Statement
May be corrosive to metals.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO â Research Use Only