Please login to your online account to display your discounted pricing

Hydrogen hexachloroplatinate(IV) hexahydrate, 99.9%, (trace metal basis), 38 to 40%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC363590250

 View more versions of this product

Catalog No. AC363590250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Trace Metal Basis
Physical Form Powder or Chunks
Formula Weight 517.92
Quantity 25g
Color Orange to orange-brown
Assay 38 to 40% (Pt)
Merck Index 14, 7526
Chemical Name or Material Hydrogen hexachloroplatinate(IV) hexahydrate
Name Note 38 to 40% Pt
Packaging Glass bottle
Assay Percent Range 99.9%
Molecular Formula H2Cl6Pt·6H2O
MDL Number MFCD00149910
CAS 18497-13-7
Total Trace Metal Impurities 0.1% max.
Synonym Hexachloroplatinic acid hexahydrate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Hydrogen hexachloroplatinate(IV) hexahydrate
Name Note 38 to 40% Pt
CAS 18497-13-7
Purity Grade Trace Metal Basis
Assay 38 to 40% (Pt)
Assay Percent Range 99.9%
Molecular Formula H2Cl6Pt·6H2O
Formula Weight 517.92
MDL Number MFCD00149910
Synonym Hexachloroplatinic acid hexahydrate
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

SDS & Safety Documents