missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen fluoride pyridine complex, ca 70% HF, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1711722

 View more versions of this product

Catalog No. AAL1711722



Description

Description

Hydrogen fluoride pyridine complex is used to prepare beta-fluoroamines via reaction with amino alcohols. As a fluorinating agent, is involved in the fluorination of 2,2-diary-1,3-dithiolanes. It is associated with nitrosonium tetrafluorborate to prepare gem-difluorides by reacting with ketoximes. It is also used to convert secondary and tertiary alcohols into their respective alkyl fluorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrogen fluoride pyridine complex is used to prepare beta-fluoroamines via reaction with amino alcohols. As a fluorinating agent, is involved in the fluorination of 2,2-diary-1,3-dithiolanes. It is associated with nitrosonium tetrafluorborate to prepare gem-difluorides by reacting with ketoximes. It is also used to convert secondary and tertiary alcohols into their respective alkyl fluorides.

Notes
Moisture sensitive. Store in cool place. Incompatible with strong bases, alkali metals, strong oxidizing agents, metals and strong acids. It reacts violently with water.
Specifications

Specifications

Hydrogen fluoride pyridine complex
1.1
≈70% HF
MFCD00012436
UN1790
Moisture sensitive
pyridine hydrofluoride, hydrogen fluoride-pyridine, olah's reagent, pyridinium fluoride, hydrogen fluoride pyridine complex, hydrogen fluoride pyridine, hf-pyridine, pyridine, hydrofluoride 1:1, pyridinium poly hydrogen fluoride, hydrogen fluoride/pyridine
C1=CC=NC=C1.F
99.108
62778-11-4
50°C (1 mmHg)
C5H6FN
100 g
4750151
14,6821
GRJJQCWNZGRKAU-UHFFFAOYSA-N
pyridine;hydrofluoride
64774

Chemical Identifiers

62778-11-4
99.108
GRJJQCWNZGRKAU-UHFFFAOYSA-N
64774
C1=CC=NC=C1.F
C5H6FN
MFCD00012436
pyridine hydrofluoride, hydrogen fluoride-pyridine, olah's reagent, pyridinium fluoride, hydrogen fluoride pyridine complex, hydrogen fluoride pyridine, hf-pyridine, pyridine, hydrofluoride 1:1, pyridinium poly hydrogen fluoride, hydrogen fluoride/pyridine
pyridine;hydrofluoride
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300+H310+H330-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: HYDROFLUORIC ACID

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only