missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen fluoride-pyridine, 65-70%, ACROS Organics™

Manufacturer:  Acros Organics 180071000

 View more versions of this product

Catalog No. AC180071000


 

Description

Description

C-X bond formation reagent
Specifications

Specifications

Hydrogen fluoride-pyridine
Yellow through Brown
65-70%
65-70%
C5H6FN
16,286; 16,050; 05,538; 06,473; 09,399; 12,419
1.1000g/mL
C1=CC=NC=C1.F
99.108
99.11
Plastic Bottle in Tin Can
1.1
100g
32001-55-1
65-70%
MFCD00012436
Pyridinium poly(hydrogen fluoride), Olah';s reagent
GRJJQCWNZGRKAU-UHFFFAOYSA-N
pyridine;hydrofluoride
64774
Liquid

Chemical Identifiers

32001-55-1
99.108
GRJJQCWNZGRKAU-UHFFFAOYSA-N
64774
C5H6FN
MFCD00012436
Pyridinium poly(hydrogen fluoride), Olah';s reagent
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger

EINECSNumber : 250-889-6

TSCA : TSCA

SDS Documents