missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Hydrogen chloride solution, 4.0 M in 1,4-dioxane, Honeywell Fluka™

4.0 M in 1,4-dioxane

$2,645.53 - $2,645.53

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.458
MDL Number MFCD00011324
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name chlorane
SMILES Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046366 View Documents Honeywell Fluka™
3455472L
2L Glass Bottle Each for $2,645.53
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7647-01-0
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane
ClH
MFCD00011324
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl

Specifications

7647-01-0
MFCD00011324
1.05g/mL at 25°C
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.46g/mol
Liquid
17°C (62.6°F)
ClH
UN2924
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.458
CHEBI:17883
Glass Bottle
2L
Hydrogen chloride solution
Safety and Handling

Safety and Handling

P210-P280-P305 + P351 + P338-P370 + P378-P403 + P235

DOTInformation : 8(3)/PGII