missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrogen chloride, nominally 2.5M in ethanol

Manufacturer:  Alfa Aesar™ 44605AK

 View more versions of this product

Catalog No. AA44605AK



Specifications

Specifications

Hydrogen chloride
ClH
UN2924
acide chlorhydrique
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.46
250mL
7647-01-0
MFCD00011324
Nominally 2.5M in ethanol
Miscible with water at room temperature. Also miscible with methanol,ethanol and ether.
Cl
36.458
CHEBI:17883
Liquid

Chemical Identifiers

7647-01-0
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane
ClH
MFCD00011324
acide chlorhydrique
CHEBI:17883
Cl
Safety and Handling

Safety and Handling

GHS H Statement
H225-H331-H314
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.

P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c

H225-H290-H314-H332-H335

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

EINECSNumber : 231-595-7

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents