Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrogen chloride, 2M in diethyl ether

$74.45 - $74.45

Chemical Identifiers

Chemical Name or Material Hydrogen chloride
CAS 7647-01-0
Molecular Formula HCl
Formula Weight 36.46
MDL Number MFCD00011324
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26914AE

  SDS
alfa aesar™
H26914AE
100mL Each for $74.45

AAH26914AP

  SDS
alfa aesar™
H26914AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 36.46
Chemical Name or Material Hydrogen chloride
TSCA TSCA
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 100mL
Health Hazard 2 GHS H Statement
H225-H331-H314-H302-H336
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Molecular Formula HCl
UN Number UN2924
Concentration 2M in diethyl ether
MDL Number MFCD00011324
Flash Point −40°C (−40°F)
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
CAS 7647-01-0
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H331-H314-H302-H336
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.

2° to 8°C