missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrogen chloride, 1M in acetic acid

$32.77 - $32.77

Chemical Identifiers

CAS 7647-01-0
Molecular Formula HCl
Formula Weight 36.46
MDL Number MFCD00011324
UN Number UN2920
Chemical Name or Material Hydrogen chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH27347AE SDS Alfa Aesar™
H27347AE
100mL Each for $32.77
AAH27347AU SDS Alfa Aesar™
H27347AU
1000mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7647-01-0
36.46
UN2920
HCl
MFCD00011324
Hydrogen chloride

Specifications

7647-01-0
1M in acetic acid
MFCD00011324
Acrid
40°C (104°F)
HCl
36.46
UN2920
100mL
Hydrogen chloride

Safety and Handling

GHS H Statement
H314-H318-H226-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

TSCA : TSCA