Please login to your online account to display your discounted pricing

Hydrogen chloride, 1.25M solution in ethanol, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC427018000

 View more versions of this product

Catalog No. AC427018000

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Hydrogen chloride
Quantity 800mL
Name Note 1.25M solution in ethanol
CAS 7647-01-0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Toxic if inhaled.
CAS 64-17-5
Health Hazard 3 GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula HCl
Linear Formula HCl
Formula Weight 36.46
MDL Number MFCD00011324
Merck Index 14, 4794
Density 0.8250g/mL
Packaging AcroSeal™ Glass Bottle

Specifications

Chemical Name or Material Hydrogen chloride
Name Note 1.25M solution in ethanol
CAS 7647-01-0
CAS 64-17-5
Molecular Formula HCl
Linear Formula HCl
Formula Weight 36.46
MDL Number MFCD00011324
Accessories

GHS Signal Word: Danger

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Toxic if inhaled.