Please login to your online account to display your discounted pricing

Hydrogen bromide, pure, 33 wt% solution in glacial acetic acid, ACROS Organics™


Quantity 500mL
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  123185000

 View more versions of this product

Catalog No. AC123185000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Fuming Solution
Formula Weight 80.9
Color Clear yellow to golden
Quantity 500mL
UN Number 2922
Purity Grade Pure
Assay 31 to 34 wt% (HBr)
Density 1.4200g/mL
Fieser 01,450; 02,214; 03,154; 04,249; 05,332; 06,282; 10,200; 15,273
Merck Index 09, 4665
Chemical Name or Material Hydrogen bromide
Water 0.5% max.
Name Note Pure
CAS 10035-10-6
MDL Number MFCD00011323
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Flammable liquid and vapor. May cause respiratory irritation. Causes severe skin burns and eye damage.
Molecular Formula HBr
Linear Formula HBr
CAS 37348-16-6
CAS 64-19-7
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes
Additional Information Vapor Pressure: 300mmHg at 20°C
Packaging Glass Bottle

Specifications

Chemical Name or Material Hydrogen bromide
Name Note Pure
CAS 37348-16-6
CAS 64-19-7
CAS 10035-10-6
Purity Grade Pure
Assay 31 to 34 wt% (HBr)
Molecular Formula HBr
Linear Formula HBr
Formula Weight 80.9
MDL Number MFCD00011323
Accessories

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes

GHS P Statement Flammable liquid and vapor. May cause respiratory irritation. Causes severe skin burns and eye damage.

SDS & Safety Documents